4-(Pyrrolidin-3-yl)morpholine ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-(3-Pyrrolidinyl)morpholine
133720-1G
133720-250MG
BT133720-1GEA
301
USD
BT133720-1G
BT133720-250MG
4-(Pyrrolidin-3-yl)morpholine ≥97%
4-(Pyrrolidin-3-yl)morpholine
CAS: 53617-37-1; MDL No: MFCD06740352
UN No: UN2735; Haz Class: 8; Packing Group: III
Solid; Molecular Formula: C8H16N2O; MW: 156.23
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₈H₁₆N₂O |
MDL Number:
MFCD06740352 CAS Number: 53617-37-1 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...