Ethyl bromodifluoroacetate ≥97%

Supplier: BeanTown Chemical

133690-1G 133690-25G 133690-5G
BT133690-1GEA 23 USD
BT133690-1G BT133690-25G BT133690-5G
Ethyl bromodifluoroacetate ≥97%
Ethyl bromodifluoroacetate

CAS: 667-27-6; EC No: 211-567-0; MDL No: MFCD00042069 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Linear Formula: BrCF2CO2CH2CH3; Molecular Formula: C4H5BrF2O2; MW: 202.99 Boiling Point: 112°/700 mmHg; Flash point: 21°C (70°F) Density (g/mL): 1.583; Refractive Index: 1.387


  • Refractive index: 1.387
  • Liquid

Formula: BrF₂CCOOCH₂CH₃
Boiling Pt: 112 °C/700 mmHg
Density: 1.583 g/mL
Flash Pt: 21 °C (70 °F)
MDL Number: MFCD00042069
CAS Number: 667-27-6
EINECS: 211-567-0
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR