1-Bromobutanone ≥85% stabilized, Technical Grade

Supplier: BeanTown Chemical

Synonyms: 1-Bromo-2-butanone

133610-1G 133610-5G
BT133610-1GEA 65 USD
BT133610-1G BT133610-5G
1-Bromobutanone ≥85% stabilized, Technical Grade
1-Bromobutanone

CAS: 816-40-0; EC No: 212-431-3; MDL No: MFCD00000207; RTECS: EL7000000 UN No: UN2927; Haz Class: 6.1 (8); Packing Group: II Liquid; Molecular Formula: C4H7BrO; MW: 151.00 Boiling Point: 105°/150 mmHg; Flash point: 68°C (154°F) Density (g/mL): 1.479; Refractive Index: 1.465


  • RTECS: EL7000000
  • Refractive index: 1.465
  • Liquid

Formula: BrH₂CC(O)CH₂CH₃
MW: 151 g/mol
Boiling Pt: 105 °C/150 mmHg
Density: 1.479 g/mL
Flash Pt: 68 °C (154 °F)
MDL Number: MFCD00000207
CAS Number: 816-40-0
EINECS: 212-431-3
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Assay 85%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR