Acetic acid glacial ≥99%

Supplier: BeanTown Chemical

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

133600-1L 133600-250ML 133600-4L
BT133600-1LEA 57.13 USD
BT133600-1L BT133600-250ML BT133600-4L
Acetic acid glacial ≥99%
Acetic acid

CAS: 64-19-7; EC No: 200-580-7; MDL No: MFCD00036152 UN No: UN2789; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: CH3CO2H; MW: 60.05 Melting Point: 16.2°; Boiling Point: 117-118°; Flash point: 40°C (104°F) Density (g/mL): 1.049; Refractive Index: 1.371


  • Refractive index: 1.371
  • Liquid

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 117…118 °C
Melting Pt: 16.2 °C
Density: 1.049 g/mL
Flash Pt: 40 °C (104 °F)
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR