Vanadium carbide 99
Supplier: BeanTown Chemical
133585-100G
133585-25G
BT133585-100GEA
145
USD
BT133585-100G
BT133585-25G
Vanadium carbide 99
Vanadium carbide
CAS: 12070-10-9; EC No: 235-122-5; MDL No: MFCD00049698
UN No: UN3178; Haz Class: 4.1; Packing Group: III
-325 Mesh Powder; Linear Formula: VC; MW: 62.95
Melting Point: 2810°
Density (g/mL): 5.77
Air Sensitive, Heat Sensitive, Moisture Sensitive
- Air Sensitive, Heat Sensitive, Moisture Sensitive
- -325 Mesh Powder
Formula:
CV MW: 62.95 Boiling Pt: 3900 Melting Pt: 2810 Density: 5.77 g/mL |
MDL Number:
MFCD00049698 CAS Number: 12070-10-9 EINECS: 235-122-5 UN: 3178 ADR: 4.1 III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...