Phosphoryl tribromide 98

Supplier: BeanTown Chemical

Synonyms: Phosphorus(V) oxybromide, Phosphorus (V) oxide bromide, Phosphoryl bromide, Phosphorus oxybromide

133575-100G 133575-25G 133575-5G
BT133575-100GEA 355.91 USD
BT133575-100G BT133575-25G BT133575-5G
Phosphoryl tribromide 98
Phosphoryl tribromide

CAS: 7789-59-5; EC No: 232-177-7; MDL No: MFCD00036298 UN No: UN1939; Haz Class: 8; Packing Group: II Solid; Linear Formula: POBr3; MW: 286.69 Melting Point: 56°; Boiling Point: 192° Density (g/mL): 2.822 Moisture Sensitive


  • Moisture Sensitive
  • Solid

Formula: Br₃OP
MW: 286.69 g/mol
Boiling Pt: 193
Melting Pt: 56 °C
Density: 2.822 g/mL
MDL Number: MFCD00036298
CAS Number: 7789-59-5
EINECS: 232-177-7
UN: 1939
ADR: 8 II
Merck Index: 12,07505

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR