Lead(IV) fluoride ≥99%

Supplier: BeanTown Chemical

Synonyms: Lead tetrafluoride

133530-1G 133530-5G
BT133530-1GEA 44 USD
BT133530-1G BT133530-5G
Lead(IV) fluoride ≥99%
Lead(IV) fluoride

CAS: 7783-59-7; EC No: 232-012-9; MDL No: MFCD00016278 UN No: UN2291; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: PbF4; MW: 283.18 Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: F₄Pb
MW: 283.19
Melting Pt: 600
Density: 6.7
MDL Number: MFCD00016278
CAS Number: 7783-59-7
EINECS: 232-012-9
UN: 2291
ADR: 6.1 III
Merck Index: 12,05448

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR