Barium chloride dihydrate ≥99%

Supplier: BeanTown Chemical

133450-1KG 133450-250G 133450-5KG
BT133450-1KGEA 80.25 USD
BT133450-1KG BT133450-250G BT133450-5KG
Barium chloride dihydrate ≥99%
Barium chloride dihydrate

CAS: 10326-27-9; EC No: 233-788-1; MDL No: MFCD00149154; RTECS: CQ8751000 UN No: UN1564; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: BaCl2·2H2O; MW: 244.26


  • RTECS: CQ8751000
  • Powder

Formula: BaCl₂·2H₂O
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR