Tetraammineplatinum(II) nitrate 99,99

Supplier: BeanTown Chemical

133430-1G 133430-250MG 133430-5G
BT133430-1GEA 200.4 USD
BT133430-1G BT133430-250MG BT133430-5G
Tetraammineplatinum(II) nitrate 99,99
Tetraammineplatinum(II) nitrate

CAS: 20634-12-2; EC No: 243-929-9; MDL No: MFCD00011624 UN No: UN1477; Haz Class: 5.1; Packing Group: III Crystalline; Linear Formula: Pt(NH3)4(NO3)2; Molecular Formula: H12N6O6Pt; MW: 387.21 Melting Point: 262° (decomposes)


  • Crystalline

Formula: H₁₂N₆O₆Pt
MW: 387.216 g/mol
Melting Pt: 262 °C (dec.)
MDL Number: MFCD00011624
CAS Number: 20634-12-2
EINECS: 243-929-9
ADR: 5.1 III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR