Arsenic(III) sulphide ≥99.999%
Supplier: BeanTown Chemical
Synonyms:
Arsenic trisulphide, Disrsenic trisulphide
133405-25G
133405-5G
BT133405-25GEA
660
USD
BT133405-25G
BT133405-5G
Arsenic(III) sulphide ≥99.999%
Arsenic(III) sulphide
CAS: 1303-33-9; EC No: 215-117-4; MDL No: MFCD00003435; RTECS: CG2638000
UN No: UN1557; Haz Class: 6.1; Packing Group: III
-100 Mesh Powder ; Molecular Formula: As2S3; MW: 246.03
- RTECS: CG2638000
- -100 Mesh Powder
Formula:
As₂S₃ MW: 246.04 g/mol Boiling Pt: 707 Melting Pt: 300 |
MDL Number:
MFCD00003435 CAS Number: 1303-33-9 EINECS: 215-117-4 UN: 1557 ADR: 6.1 III Merck Index: 12,00846 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...