3-Bromothiophene ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-Thienyl bromide

133400-10G 133400-250G 133400-50G
BT133400-10GEA 32 USD
BT133400-10G BT133400-250G BT133400-50G
3-Bromothiophene ≥97%
3-Bromothiophene

CAS: 872-31-1; EC No: 212-821-3; MDL No: MFCD00005464 UN No: UN2810; Haz Class: 6.1; Packing Group: II Liquid; Molecular Formula: C4H3BrS; MW: 163.04 Boiling Point: 150°; Flash point: 52°C (125°F) Density (g/mL): 1.74; Refractive Index: 1.591


  • Refractive index: 1.591
  • Liquid

Formula: C₄H₃BrS
MW: 163.04 g/mol
Boiling Pt: 150 °C
Density: 1.74 g/mL
Flash Pt: 52 °C (125 °F)
MDL Number: MFCD00005464
CAS Number: 872-31-1
EINECS: 212-821-3
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR