1,1,3,3-Tetramethyldisiloxane ≥97%

Supplier: BeanTown Chemical

133355-100G 133355-25G 133355-500G
BT133355-100GEA 110 USD
BT133355-100G BT133355-25G BT133355-500G
1,1,3,3-Tetramethyldisiloxane ≥97%
1,1,3,3-Tetramethyldisiloxane

CAS: 3277-26-7; EC No: 221-906-4; MDL No: MFCD00008256; RTECS: JN1000000 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4H14OSi2 ; MW: 134.32 Boiling Point: 70-71°; Flash point: -26°C (-15°F) Density (g/mL): 0.760 ; Refractive Index: 1.370 Moisture Sensitive


  • RTECS: JN1000000
  • Refractive index: 1.370
  • Moisture Sensitive
  • Liquid

Formula: C₄H₁₄OSi₂
MW: 134.33 g/mol
Boiling Pt: 70…71 °C
Density: 0.760 g/mL
Flash Pt: –26 °C (–15 °F)
MDL Number: MFCD00008256
CAS Number: 3277-26-7
EINECS: 221-906-4
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR