Trichloro(1H,1H,2H,2H-heptadecafluorodecyl)silane ≥96%

Supplier: BeanTown Chemical

Synonyms: 1-(Trichlorosilyl)-1H,1H,2H,2H-heptadecafluorodecane, Trichloro(1H,1H,2H,2H-perfluorodecyl)silane, 1H,1H,2H,2H-Perfluorodecyltrichlorosilane

133325-1G 133325-25G 133325-5G
BT133325-1GEA 34 USD
BT133325-1G BT133325-25G BT133325-5G
Trichloro(1H,1H,2H,2H-heptadecafluorodecyl)silane ≥96%
Trichloro(1H,1H,2H,2H-heptadecafluorodecyl)silane

CAS: 78560-44-8; MDL No: MFCD00042344 UN No: UN2987; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C10H4Cl3F17Si; MW: 581.56 Density (g/mL): 1.54 Moisture Sensitive


  • Moisture Sensitive
  • Liquid

Formula: C₁₀H₄Cl₃F₁₇Si
MW: 581.55 g/mol
Density: 1.54 g/mL
MDL Number: MFCD00042344
CAS Number: 78560-44-8
ADR: 8,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR