1-Dimethylamino-2-propyne ≥97%
Supplier: BeanTown Chemical
CAS: 7223-38-3; EC No: 230-620-9; MDL No: MFCD00008575
UN No: UN2733; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: HC≡CCH2N(CH3)2; Molecular Formula: C5H9N; MW: 83.13
Boiling Point: 79-83°; Flash point: -8°C (18°F)
Density (g/mL): 0.772; Refractive Index: 1.419
- Refractive index: 1.419
- Liquid
Formula:
C₅H₉N Boiling Pt: 79…83 °C Density: 0.772 g/mL Flash Pt: –8 °C (18 °F) |
MDL Number:
MFCD00008575 CAS Number: 7223-38-3 EINECS: 230-620-9 ADR: 3 (8),II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...