Zinc sulfate hydrate ≥99.999%

Supplier: BeanTown Chemical

133280-10G 133280-50G
BT133280-10GEA 53.33 USD
BT133280-10G BT133280-50G
Zinc sulfate hydrate ≥99.999%
Zinc sulfate hydrate

CAS: 16788-42-4; EC No: 231-793-3; MDL No: MFCD00011302 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline/Powder; Linear Formula: ZnSO4·xH2O ; MW: 161.45 (anhydrous)


  • Crystalline/Powder

Formula: ZnSO₄·xH₂O
MW: 161.45 g/mol (anhy)
MDL Number: MFCD00149894
CAS Number: 16788-42-4
EINECS: 231-793-3
ADR: 9 III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR