Manganese(II) oxalate dihydrate

Supplier: BeanTown Chemical

133250-250G 133250-50G
BT133250-250GEA 213 USD
BT133250-250G BT133250-50G
Manganese(II) oxalate dihydrate
Manganese(II) oxalate dihydrate

CAS: 6556-16-7; EC No: 211-367-3; MDL No: MFCD00150452 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder/Chunks; Linear Formula: MnC2O4·2H2O; MW: 178.99 Melting Point: 100° (decomposes) Density (g/mL): 2.453


  • Powder/Chunks

Formula: C₂H₆MnO₆
MW: 181.002 g/mol
Melting Pt: 100 °C (dec.)
Density: 2.453 g/mL
MDL Number: MFCD00150452
CAS Number: 6556-16-7
EINECS: 211-367-3
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR