2,6-Dimethoxyaniline ≥97%

Supplier: BeanTown Chemical

133195-1G 133195-5G
BT133195-1GEA 99 USD
BT133195-1G BT133195-5G
2,6-Dimethoxyaniline ≥97%
2,6-Dimethoxyaniline

CAS: 2734-70-5; MDL No: MFCD00053934 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Molecular Formula: C8H11NO2; MW: 153.18


  • Crystalline/Powder

Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
MDL Number: MFCD00053934
CAS Number: 2734-70-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR