2-(Trifluoromethoxy)benzoyl chloride ≥97%

Supplier: BeanTown Chemical

133190-1G 133190-25G 133190-5G
BT133190-1GEA 48 USD
BT133190-1G BT133190-25G BT133190-5G
2-(Trifluoromethoxy)benzoyl chloride ≥97%
2-(Trifluoromethoxy)benzoyl chloride

CAS: 162046-61-9; MDL No: MFCD00052330 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C8H4ClF3O2 ; MW: 224.56 Density (g/mL): 1.435 Moisture Sensitive


  • Moisture Sensitive
  • Liquid

Formula: C₈H₄ClF₃O₂
Density: 1.435 g/mL
MDL Number: MFCD00052330
CAS Number: 162046-61-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR