2-Chloroethyl isocyanate ≥97%

Supplier: BeanTown Chemical

Synonyms: Isocyanic acid 2-Chloroethyl ester

133145-100G 133145-25G 133145-5G
BT133145-100GEA 244 USD
BT133145-100G BT133145-25G BT133145-5G
2-Chloroethyl isocyanate ≥97%
2-Chloroethyl isocyanate

CAS: 1943-83-5; EC No: 217-734-4; MDL No: MFCD00002043; RTECS: NQ8450000 UN No: UN3080; Haz Class: 6.1 (3); Packing Group: II Liquid; Linear Formula: ClCH2CH2NCO ; Molecular Formula: C3H4ClNO; MW: 105.52 Boiling Point: 141-142°; Flash point: 56°C (133°F) Density (g/mL): 1.237; Refractive Index: 1.447 Moisture Sensitive


  • RTECS: NQ8450000
  • Refractive index: 1.447
  • Moisture Sensitive
  • Liquid

Formula: C₃H₄ClNO
MW: 105.52 g/mol
Boiling Pt: 141…142 °C
Density: 1.237 g/mL
Flash Pt: 56 °C (133 °F)
MDL Number: MFCD00002043
CAS Number: 1943-83-5
EINECS: 217-734-4
UN: 3080
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR