Sodium isopropoxide
Supplier: BeanTown Chemical
133065-100G
133065-25G
133065-5G
BT133065-100GEA
336.99
USD
BT133065-100G
BT133065-25G
BT133065-5G
Sodium isopropoxide
Sodium isopropoxide
CAS: 683-60-3; EC No: 211-673-7; MDL No: MFCD00061566
UN No: UN3206; Haz Class: 4.2 (8); Packing Group: II
Crystalline; Molecular Formula: C3H7NaO; MW: 82.08
Melting Point: 70-75°
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Crystalline
Formula:
C₃H₇NaO Melting Pt: 70…75 °C |
MDL Number:
MFCD00061566 CAS Number: 683-60-3 EINECS: 211-673-7 ADR: 4.2 (8),II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...