Dibutyl phthalate ≥99%
Supplier: BeanTown Chemical
Synonyms:
DBP, Phthalic acid dibutyl ester
133060-1KG
133060-25G
133060-5G
BT133060-1KGEA
57.79
USD
BT133060-1KG
BT133060-25G
BT133060-5G
Dibutyl phthalate ≥99%
Dibutyl phthalate
CAS: 84-74-2; EC No: 201-557-4; MDL No: MFCD00009441; RTECS: TI0875000
UN No: UN3082; Haz Class: 9; Packing Group: III
Liquid; Molecular Formula: C16H22O4; MW: 278.35
Melting Point: -35°; Boiling Point: 340°; Flash point: 171°C (340°F)
Density (g/mL): 1.043; Refractive Index: 1.492
- RTECS: TI0875000
- Refractive index: 1.492
- Liquid
Formula:
C₁₆H₂₂O₄ MW: 278.35 g/mol Boiling Pt: 340 °C Melting Pt: –35 °C Density: 1.043 g/mL Flash Pt: 171 °C (340 °F) |
MDL Number:
MFCD00009441 CAS Number: 84-74-2 EINECS: 201-557-4 UN: 3082 ADR: 9,III |
|
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...