2,4,6-Trimethoxyboroxin
Supplier: BeanTown Chemical
Synonyms:
Trimethoxycyclotriboroxane, Trimethoxyboroxine
133050-250G
133050-50G
BT133050-250GEA
74
USD
BT133050-250G
BT133050-50G
2,4,6-Trimethoxyboroxin
2,4,6-Trimethoxyboroxin
CAS: 102-24-9; EC No: 203-016-8; MDL No: MFCD00005946; RTECS: ED8720000
UN No: UN2924; Haz Class: 3(8); Packing Group: II
Liquid; Linear Formula: (CH3O)3B3O3; Molecular Formula: C3H9B3O6; MW: 173.53
Melting Point: 10-11°; Boiling Point: 130°; Flash point: 9°C (48°F)
Density (g/mL): 1.195; Refractive Index: 1.400
Air Sensitive, Moisture Sensitive
- RTECS: ED8720000
- Refractive index: 1.400
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₃H₉B₃O₆ MW: 173.53 g/mol Boiling Pt: 130 °C Melting Pt: 10…11 °C Density: 1.195 g/mL Flash Pt: 9 °C (48 °F) |
MDL Number:
MFCD00005946 CAS Number: 102-24-9 EINECS: 203-016-8 ADR: 3(8),II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...