3-Fluoro-o-toluidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Amino-6-fluorotoluene, 3-Fluoro-2-methylaniline

132960-25G 132960-5G
BT132960-25GEA 245 USD
BT132960-25G BT132960-5G
3-Fluoro-o-toluidine ≥98%
3-Fluoro-o-toluidine

CAS: 443-86-7; EC No: 207-142-4; MDL No: MFCD00007760 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C7H8FN; MW: 125.14 Melting Point: 7°; Boiling Point: 89-91°/15 mmHg; Flash point: 86°C (187°F) Density (g/mL): 1.099; Refractive Index: 1.542


  • Refractive index: 1.542
  • Liquid

Formula: C₇H₈FN
MW: 125.15
Boiling Pt: 203
Melting Pt: 7
Density: 1.099 g/mL
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00007760
CAS Number: 443-86-7
EINECS: 207-142-4
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR