Aluminium chloride, anhydrous ≥99%

Supplier: BeanTown Chemical

Synonyms: Aluminum chloride

132915-100G 132915-500G
BT132915-100GEA 53.3 USD
BT132915-100G BT132915-500G
Aluminium chloride, anhydrous ≥99%
Aluminium chloride

CAS: 7446-70-0; EC No: 231-208-1; MDL No: MFCD00003422; RTECS: BD0525000 UN No: UN1726; Haz Class: 8; Packing Group: II Powder; Molecular Formula: AlCl3; MW: 133.34 Melting Point: 190° Moisture Sensitive


  • RTECS: BD0525000
  • Moisture Sensitive
  • Powder

Formula: AlCl₃
MW: 133.34 g/mol
Boiling Pt: 180
Melting Pt: 190 °C
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 13,00335

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR