Amidosulfonic acid ≥98%

Supplier: BeanTown Chemical

132905-100G 132905-2.5KG 132905-500G
BT132905-100GEA 25.57 USD
BT132905-100G BT132905-2.5KG BT132905-500G
Amidosulfonic acid ≥98%
Amidosulphonic acid

CAS: 5329-14-6; EC No: 226-218-8; MDL No: MFCD00011603; RTECS: WO5950000 UN No: UN2967; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: NH2SO3H; MW: 97.09 Melting Point: 215-225° (decomposes)


  • RTECS: WO5950000
  • Crystalline

Formula: H₃NO₃S
Melting Pt: 215…225 °C (dec.)
MDL Number: MFCD00011603
CAS Number: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR