Lanthanum(III) nitrate hydrate ≥99%
Supplier: BeanTown Chemical
132855-100G
132855-500G
BT132855-100GEA
50.92
USD
BT132855-100G
BT132855-500G
Lanthanum(III) nitrate hydrate ≥99%
Lanthanum(III) nitrate hydrate
CAS: 100587-94-8; EC No: 233-238-0; MDL No: MFCD00011067; RTECS: OE5075000
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: LaN3O9·xH2O; MW: 324.92 (anhydrous)
Melting Point: 65-68°
- RTECS: OE5075000
- Crystalline
Formula:
H₂LaN₃O₁₀ MW: 342.932 g/mol |
MDL Number:
MFCD00011067 CAS Number: 100587-94-8 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...