Tetrahydrofuran ≥99% stabilised

Supplier: BeanTown Chemical

Synonyms: Butylene oxide, THF, Diethylene oxide

132850-1L 132850-2.5L 132850-500ML
BT132850-1LEA 55.95 USD
BT132850-1L BT132850-2.5L BT132850-500ML
Tetrahydrofuran ≥99% stabilised
Tetrahydrofuran

CAS: 109-99-9; EC No: 203-726-8; MDL No: MFCD00005356; RTECS: LU5950000 UN No: UN2056; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4H8O ; MW: 72.11 Melting Point: -108°; Boiling Point: 65-67°; Flash point: -17°C (1°F) Density (g/mL): 0.889; Refractive Index: 1.407


  • RTECS: LU5950000
  • Refractive index: 1.407
  • Liquid

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: -108.5
Density: 0.889 g/mL
Flash Pt: –17 °C (1 °F)
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR