3-Phenyl-2-propynenitrile ≥98%
Supplier: BeanTown Chemical
CAS: 935-02-4; MDL No: MFCD19300770; RTECS: UE0220000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline/Lump; Molecular Formula: C9H5N; MW: 127.14
Melting Point: 37-42°; Boiling Point: 89°/3 mmHg
Refractive Index: 1.005
- RTECS: UE0220000
- Refractive index: 1.005
- Crystalline/Lump
Formula:
C₉H₅N Boiling Pt: 89 °C/3 mmHg Melting Pt: 37…42 °C |
MDL Number:
MFCD19300770 CAS Number: 935-02-4 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...