2-(Difluoromethoxy)benzylamine ≥97%
Supplier: BeanTown Chemical
Synonyms:
(2-(Difluoromethoxy)phenyl)methanamine
132810-1G
132810-250MG
BT132810-1GEA
135
USD
BT132810-1G
BT132810-250MG
2-(Difluoromethoxy)benzylamine ≥97%
2-(Difluoromethoxy)benzylamine
CAS: 243863-36-7; MDL No: MFCD00236229
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C8H9F2NO; MW: 173.16
Air Sensitive
- Air Sensitive
- Liquid
Formula:
C₈H₉F₂NO MW: 173.16 g/mol |
MDL Number:
MFCD00236229 CAS Number: 243863-36-7 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...