(2-Chlorophenyl)acetonitrile ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Chlorobenzyl cyanide, 2-(2-Chlorophenyl)acetonitrile

132635-10G 132635-50G
BT132635-10GEA 27 USD
BT132635-10G BT132635-50G
(2-Chlorophenyl)acetonitrile ≥99%
(2-Chlorophenyl)acetonitrile

CAS: 2856-63-5; EC No: 220-669-4; MDL No: MFCD00001898 UN No: UN2811 ; Haz Class: 6.1; Packing Group: II Liquid/Low Melting Solid; Molecular Formula: C8H6ClN; MW: 151.59 Melting Point: 24°; Boiling Point: 240-242°; Flash point: 110°C (230°F) Refractive Index: 1.544


  • Refractive index: 1.544
  • Liquid/Low Melting Solid

Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 240…242 °C
Melting Pt: 24 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00001898
CAS Number: 2856-63-5
EINECS: 220-669-4
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR