2-(4-Aminophenyl)ethylamine ≥98%

Supplier: BeanTown Chemical

132625-1G 132625-5G
BT132625-1GEA 51.99 USD
BT132625-1G BT132625-5G
2-(4-Aminophenyl)ethylamine ≥98%
2-(4-Aminophenyl)ethylamine

CAS: 13472-00-9; EC No: 236-739-2; MDL No: MFCD00008194 UN No: UN3259; Haz Class: 8; Packing Group: III Liquid/Low Melting Solid; Molecular Formula: C8H12N2; MW: 136.19 Melting Point: 28-31°; Boiling Point: 103°/0.3 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.034; Refractive Index: 1.591 Air Sensitive


  • Refractive index: 1.591
  • Air Sensitive
  • Liquid/Low Melting Solid

Formula: C₈H₁₂N₂
Boiling Pt: 103 °C/0.3 mmHg
Melting Pt: 28…31 °C
Density: 1.034 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00008194
CAS Number: 13472-00-9
EINECS: 236-739-2
ADR: 8,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR