Diethyl vinylphosphonate ≥97%

Supplier: BeanTown Chemical

132585-10G 132585-2G 132585-500MG
BT132585-10GEA 207 USD
BT132585-10G BT132585-2G BT132585-500MG
Diethyl vinylphosphonate ≥97%
Diethyl vinylphosphonate

CAS: 682-30-4; EC No: 211-663-2; MDL No: MFCD00009079 UN No: UN3278; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: CH2=CHPO(OCH2CH3)2 ; Molecular Formula: C6H13O3P; MW: 164.14 Boiling Point: 202°; Flash point: 110°C (230°F) Density (g/mL): 1.068 ; Refractive Index: 1.429


  • Refractive index: 1.429
  • Liquid

Formula: C₆H₁₃O₃P
Boiling Pt: 202 °C
Density: 1.068 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00009079
CAS Number: 682-30-4
EINECS: 211-663-2
UN: 3278
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR