1-Hexyne ≥98%
Supplier: BeanTown Chemical
CAS: 693-02-7; EC No: 211-736-9; MDL No: MFCD00009504; RTECS: MQ9691000
UN No: UN3295; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3(CH2)3C≡CH; Molecular Formula: C6H10; MW: 82.14
Melting Point: -132°; Boiling Point: 71-72°; Flash point: -20°C (-4°F)
Density (g/mL): 0.715; Refractive Index: 1.399
- RTECS: MQ9691000
- Refractive index: 1.399
- Liquid
Formula:
C₆H₁₀ Boiling Pt: 71…72 °C Melting Pt: –132 °C Density: 0.715 g/mL Flash Pt: –20 °C (–4 °F) |
MDL Number:
MFCD00009504 CAS Number: 693-02-7 EINECS: 211-736-9 UN: 3295 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...