Sodium selenide ≥99.5%

Supplier: BeanTown Chemical

132575-1G 132575-5G
BT132575-1GEA 306 USD
BT132575-1G BT132575-5G
Sodium selenide ≥99.5%
Sodium selenide

CAS: 1313-85-5; EC No: 215-212-0; MDL No: MFCD00014240; RTECS: WE0350000 UN No: UN3283; Haz Class: 6.1; Packing Group: II -60 Mesh Powder; Molecular Formula: Na2Se ; MW: 124.94 Melting Point: 875° Density (g/mL): 2.625 Air Sensitive, Hygroscopic, Moisture Sensitive


  • RTECS: WE0350000
  • Air Sensitive, Hygroscopic, Moisture Sensitive
  • -60 Mesh Powder

Formula: Na₂Se
MW: 124.94
Melting Pt: >875 °C
Density: 2.625 g/mL
MDL Number: MFCD00014240
CAS Number: 1313-85-5
EINECS: 215-212-0
UN: 3283
ADR: 6.1 II
Merck Index: 12,08821

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR