1,4-Dioxane, anhydrous ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
132540-100ML
132540-1L
132540-4X1L
BT132540-100MLEA
59.39
USD
BT132540-100ML
BT132540-1L
BT132540-4X1L
1,4-Dioxane, anhydrous ≥99.5%
1,4-Dioxane
CAS: 123-91-1; EC No: 204-661-8; MDL No: MFCD00006571; RTECS: JG8225000
UN No: UN1165; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C4H8O2; MW: 88.11
Melting Point: 10-12°; Boiling Point: 100-102°; Flash point: 12°C (54°F)
Density (g/mL): 1.034; Refractive Index: 1.422
Hygroscopic
- RTECS: JG8225000
- Refractive index: 1.422
- Hygroscopic
- Liquid
Formula:
C₄H₈O₂ MW: 88.11 g/mol Boiling Pt: 100…102 °C Melting Pt: 10…12 °C Density: 1.034 g/mL Flash Pt: 12 °C (54 °F) |
MDL Number:
MFCD00006571 CAS Number: 123-91-1 EINECS: 204-661-8 UN: 1165 ADR: 3 II Merck Index: 13,03330 |
|
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...