Tin(II) chloride dihydrate
Supplier: BeanTown Chemical
Synonyms:
Stannous chloride dihydrate
132455-100G
132455-500G
BT132455-100GEA
32.99
USD
BT132455-100G
BT132455-500G
Tin(II) chloride dihydrate
Tin (II) chloride dihydrate
CAS: 10025-69-1; EC No: 231-868-0; MDL No: MFCD00149863; RTECS: XP8850000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: SnCl2·2H2O; MW: 225.65
Melting Point: 37-38° (decomposes); Boiling Point: 652°
Density (g/mL): 2.71
Air Sensitive, Moisture Sensitive
- RTECS: XP8850000
- Air Sensitive, Moisture Sensitive
- Crystalline
Formula:
SnCl₂·2H₂O MW: 225.65 g/mol Boiling Pt: 652 °C Melting Pt: 37…38 °C (dec.) Density: 2.71 g/mL |
MDL Number:
MFCD00149863 CAS Number: 10025-69-1 EINECS: 231-868-0 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...