Sodium aluminate, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Sodium Aluminium oxide, Aluminum sodium oxide, Aluminium sodium dioxide, Sodium aluminum oxide

132435-100G 132435-500G
BT132435-100GEA 63.96 USD
BT132435-100G BT132435-500G
Sodium aluminate, Technical Grade
Sodium aluminate

CAS: 1302-42-7; EC No: 215-100-1; MDL No: MFCD00801010 UN No: UN2812; Haz Class: 8; Packing Group: III Crystalline/Powder; Molecular Formula: Al2O3·Na2O; MW: 163.94 Melting Point: 1650° Density (g/mL): 2.6 Hygroscopic


  • Hygroscopic
  • Crystalline/Powder

Formula: NaAlO₂
MW: 81.97 g/mol
Melting Pt: 1650 °C
Density: 2.6 g/mL
MDL Number: MFCD00801010
CAS Number: 1302-42-7
EINECS: 215-100-1
UN: 2812
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR