(±)-3-Amino-1,2-propanediol ≥97%

Supplier: BeanTown Chemical

132430-100G 132430-25G 132430-500G
BT132430-100GEA 171.16 USD
BT132430-100G BT132430-25G BT132430-500G
(±)-3-Amino-1,2-propanediol ≥97%
(±)-3-Amino-1,2-propanediol

CAS: 616-30-8; EC No: 210-475-8; MDL No: MFCD00008140; RTECS: TY2800000 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Linear Formula: NH2CH2CH(OH)CH2OH; Molecular Formula: C3H9NO2 ; MW: 91.11 Boiling Point: 264-265°/739 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.175; Refractive Index: 1.492 Hygroscopic


  • RTECS: TY2800000
  • Refractive index: 1.492
  • Hygroscopic
  • Liquid

Formula: C₃H₉NO₂
Boiling Pt: 264…265 °C/739 mmHg
Density: 1.175 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00008140
CAS Number: 616-30-8
EINECS: 210-475-8
ADR: 8,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR