Dibutyl phosphate ≥96%
Supplier: BeanTown Chemical
Synonyms:
Dibutylphosphate, Phosphoric Acid Dibutyl Ester
132400-1L
132400-250ML
BT132400-1LEA
165
USD
BT132400-1L
BT132400-250ML
Dibutyl phosphate ≥96%
Dibutyl phosphate
CAS: 107-66-4; EC No: 203-509-8; MDL No: MFCD00015245; RTECS: TB9605000
UN No: UN1718; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: (CH3(CH2)3O)2P(O)OH; Molecular Formula: C8H19; MW: 210.21
Boiling Point: 250°; Flash point: 157°C (315°F)
Density (g/mL): 1.06
- RTECS: TB9605000
- Liquid
Formula:
C₈H₁₉O₄P MW: 210.21 g/mol Boiling Pt: 250 °C Density: 1.06 g/mL Flash Pt: 157 °C (315 °F) |
MDL Number:
MFCD00015245 CAS Number: 107-66-4 EINECS: 203-509-8 ADR: 8 III |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...