Trimethylsilylacetylene ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethynyltrimethylsilane

132320-100G 132320-25G 132320-5G
BT132320-100GEA 439 USD
BT132320-100G BT132320-25G BT132320-5G
Trimethylsilylacetylene ≥98%
Trimethylsilylacetylene

CAS: 1066-54-2; EC No: 213-919-9; MDL No: MFCD00008569 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: HC≡CSi(CH3)3; Molecular Formula: C5H10Si; MW: 98.22 Boiling Point: 53°; Flash point: -34°C (-29°F) Density (g/mL): 0.695; Refractive Index: 1.388


  • Refractive index: 1.388
  • Liquid

Formula: C₅H₁₀Si
MW: 98.22 g/mol
Boiling Pt: 53 °C
Density: 0.695 g/mL
Flash Pt: –34 °C (–29 °F)
MDL Number: MFCD00008569
CAS Number: 1066-54-2
EINECS: 213-919-9
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR