Gallium indium alloy (75.5:24.5 w%), eutectic ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Gallium-indium eutectic
132295-10G
132295-50G
BT132295-10GEA
157.8
USD
BT132295-10G
BT132295-50G
Gallium indium alloy (75.5:24.5 w%), eutectic ≥99.99% (trace metals basis)
Gallium Indium alloy (75.5:24.5 w%), eutectic
EC No: 231-163-8; MDL No: MFCD00144387
UN No: UN1760; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: Ga:In; 75.5:24.5 wt%
Melting Point: 15.7°
Density (g/mL): 6.25
- Liquid
Melting Pt:
15.7 °C Density: 6.25 g/mL |
MDL Number:
MFCD00144387 CAS Number: 56451-02-6 EINECS: 231-163-8 ADR: 8,III |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...