1,3-Difluoro-4-nitrobenzene ≥98%
Supplier: BeanTown Chemical
Synonyms:
2,4-Difluoro-1-nitrobenzene, 2,4-Difluoronitrobenzene
132215-100G
132215-25G
132215-500G
BT132215-100GEA
95
USD
BT132215-100G
BT132215-25G
BT132215-500G
1,3-Difluoro-4-nitrobenzene ≥98%
1,3-Difluoro-4-nitrobenzene
CAS: 446-35-5; EC No: 207-167-0; MDL No: MFCD00007050; RTECS: CZ5730000
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H3F2NO2; MW: 159.09
Melting Point: 9-10°; Boiling Point: 203-204°; Flash point: 91°C (196°F)
Density (g/mL): 1.451; Refractive Index: 1.511
- RTECS: CZ5730000
- Refractive index: 1.511
- Liquid
Formula:
C₆H₃F₂NO₂ MW: 159.09 g/mol Boiling Pt: 203…204 °C Melting Pt: 9…10 °C Density: 1.451 g/mL Flash Pt: 91 °C (196 °F) |
MDL Number:
MFCD00007050 CAS Number: 446-35-5 EINECS: 207-167-0 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...