Glyoxylic acid monohydrate ≥97%

Supplier: BeanTown Chemical

Synonyms: Formylformic acid monohydrate, Oxoacetic acid monohydrate, 2-Oxoethanoic acid monohydrate

132185-100G 132185-25G
BT132185-100GEA 95 USD
BT132185-100G BT132185-25G
Glyoxylic acid monohydrate ≥97%
Glyoxylic acid monohydrate

CAS: 563-96-2; EC No: 206-058-5; MDL No: MFCD00149497; RTECS: MD4550000 UN No: UN3261; Haz Class: 8; Packing Group: III Solid; Linear Formula: HCOCO2H·H2O; Molecular Formula: C2H2O3·H2O; MW: 92.06 Melting Point: 49-52°; Boiling Point: 100°; Flash point: 110°C (230°F) Hygroscopic


  • RTECS: MD4550000
  • Hygroscopic
  • Solid

Formula: C₂H₂O₃·1H₂O
MW: 92.05 g/mol
Boiling Pt: 100 °C
Melting Pt: 49…52 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00127974
CAS Number: 563-96-2
EINECS: 206-058-5
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR