(Methyl)triphenylphosphonium iodide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Methyltriphenylphosphonium iodide
132165-100G
132165-25G
132165-500G
BT132165-100GEA
155
USD
BT132165-100G
BT132165-25G
BT132165-500G
(Methyl)triphenylphosphonium iodide ≥98%
(Methyl)triphenylphosphonium iodide
CAS: 2065-66-9; EC No: 218-178-5; MDL No: MFCD00066175
UN No: UN3464; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: CH3P(C6H5)3I; Molecular Formula: C19H18IP; MW: 404.23
Melting Point: 183-185°
Hygroscopic , Light Sensitive
- Hygroscopic, Light Sensitive
- Powder
Formula:
C₁₉H₁₈IP MW: 404.23 g/mol Melting Pt: 183…185 °C |
MDL Number:
MFCD00066175 CAS Number: 2065-66-9 EINECS: 218-178-5 UN: 3464 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...