Phosphotungstic acid hydrate
Supplier: BeanTown Chemical
132135-250G
132135-50G
BT132135-250GEA
317
USD
BT132135-250G
BT132135-50G
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
CAS: 12501-23-4; MDL No: MFCD00011342; RTECS: TH5650000
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Linear Formula: H3PO4·12WO3·xH2O; MW: 2880.16
Melting Point: 95°
Light Sensitive, Moisture Sensitive
- RTECS: TH5650000
- Light Sensitive, Moisture Sensitive
- Powder
Formula:
H₃[P(W₃O₁₀)₄]·nH₂O Melting Pt: 95 °C |
MDL Number:
MFCD00011342 CAS Number: 12501-23-4 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...