Tin(II) oxalate ≥98%

Supplier: BeanTown Chemical

132130-100G 132130-2.5KG 132130-500G
BT132130-100GEA 58 USD
BT132130-100G BT132130-2.5KG BT132130-500G
Tin(II) oxalate ≥98%
Tin(II) oxalate

CAS: 814-94-8; EC No: 212-414-0; MDL No: MFCD00040678; RTECS: XQ3950000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: SnC2O4; MW: 206.73 Melting Point: 280° (decomposes) Density (g/mL): 3.56


  • RTECS: XQ3950000
  • Powder

Formula: C₂O₄Sn
Melting Pt: 280 °C (dec.)
Density: 3.56 g/mL
MDL Number: MFCD00040678
CAS Number: 814-94-8
EINECS: 212-414-0
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR