Nickel ≥99.9% (trace metals basis), powder APS 2.2-3.0 µm

Supplier: BeanTown Chemical

132100-100G 132100-500G
BT132100-100GEA 44.96 USD
BT132100-100G BT132100-500G
Nickel ≥99.9% (trace metals basis), powder APS 2.2-3.0 µm
Nickel

CAS: 7440-02-0; EC No: 231-111-4; MDL No: MFCD00011137; RTECS: QR5950000 UN No: UN3089; Haz Class: 4.1; Packing Group: II Powder; Molecular Formula: Ni; MW: 58.69 Melting Point: 1453°; Boiling Point: 2732° Density (g/mL): 8.9


  • RTECS: QR5950000
  • Powder

Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 2732 °C
Melting Pt: 1453 °C
Density: 8.9 g/mL
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR