1-Bromo-2-butyne ≥98%
Supplier: BeanTown Chemical
132085-25G
132085-5G
BT132085-25GEA
447
USD
BT132085-25G
BT132085-5G
1-Bromo-2-butyne ≥98%
1-Bromo-2-butyne
CAS: 3355-28-0; MDL No: MFCD00190233
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3C≡CCH2Br; Molecular Formula: C4H5Br; MW: 132.99
Boiling Point: 40-41°/20 mmHg; Flash point: 36°C (97°F)
Density (g/mL): 1.519; Refractive Index: 1.508
- Refractive index: 1.508
- Liquid
Formula:
C₄H₅Br MW: 132.99 g/mol Boiling Pt: 40…41 °C/20 mmHg Density: 1.519 g/mL Flash Pt: 36 °C (97 °F) |
MDL Number:
MFCD00190233 CAS Number: 3355-28-0 UN: 1993 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...