Procaine hydrochloride ≥99%

Supplier: BeanTown Chemical

132010-250G 132010-50G
BT132010-250GEA 104 USD
BT132010-250G BT132010-50G
Procaine hydrochloride ≥99%
Procaine hydrochloride

CAS: 51-05-8; EC No: 200-077-2; MDL No: MFCD00013000; RTECS: DG2275000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C13H20N2O2·HCl ; MW: 272.77 Melting Point: 155-156° Air Sensitive


  • RTECS: DG2275000
  • Air Sensitive
  • Crystalline

Formula: C₁₃H₂₁ClN₂O₂
Melting Pt: 155…156 °C
MDL Number: MFCD00013000
CAS Number: 51-05-8
EINECS: 200-077-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR