Lead(II) oxide ≥99.9995% (trace metals basis)

Supplier: BeanTown Chemical

131975-100G 131975-25G 131975-500G
BT131975-100GEA 374.84 USD
BT131975-100G BT131975-25G BT131975-500G
Lead(II) oxide ≥99.9995% (trace metals basis)
Lead(II) oxide

CAS: 1317-36-8; EC No: 215-267-0; MDL No: MFCD00011164; RTECS: OG1750000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: PbO; MW: 223.19 Melting Point: 886°


  • RTECS: OG1750000
  • Powder

Formula: PbO
Melting Pt: 886 °C
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.9995%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR